2-Methyl-2-propanyl-(2S)-2-cyan-1-pyrrolidincarboxylat 97%

Lieferant: Apollo Scientific

Synonyme: (S)-1-Boc-2-pyrrolidinecarbonitrile, (S)-1-(tert-Butoxycarbonyl)-2-cyanopyrrolidine, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinecarbonitrile, (S)-1-Boc-2-cyanopyrrolidine, tert-Butyl (S)-2-Cyano-1-pyrrolidinecarboxylate, (S)-tert-Butyl 2-cyanopyrrolidine-1-carboxylate, (S)-2-Cyano-1-pyrrolidinecarboxyic acid tert-Butyl ester

OR4610-10G
APOSOR4610-10GEA 82.8 EUR
APOSOR4610-10G
2-Methyl-2-propanyl-(2S)-2-cyan-1-pyrrolidincarboxylat 97%
(S)-tert-Butyl-2-cyanopyrrolidine-1-carboxylate
Formel: C₁₀H₁₆N₂O₂
Molecular Weight: 196,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01861224
CAS-Nummer: 228244-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR