4-Chloro-3-iodo-1H-pyrrolo[2,3-b]pyridine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-3-iodo-7-azaindole
OR46101-5G
APOSOR46101-5GEA
234
EUR
APOSOR46101-5G
4-Chloro-3-iodo-1H-pyrrolo[2,3-b]pyridine 97%
4-Chloro-3-iodo-1H-pyrrolo[2,3-b]pyridine
Formel:
C₇H₄ClIN₂ Molecular Weight: 278,48 g/mol Schmelzpunkt: 202…204 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09266226 CAS-Nummer: 869335-73-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...