1H-Pyrrolo[2,3-b]pyridine-3-carbaldehyde 98%
Lieferant: Apollo Scientific
Synonyme:
7-Azaindole-3-carboxaldehyde
OR46102-25G
APOSOR46102-25GEA
94.4
EUR
APOSOR46102-25G
1H-Pyrrolo[2,3-b]pyridine-3-carbaldehyde 98%
1H-Pyrrolo[2,3-b]pyridine-3-carbaldehyde
Formel:
C₈H₆N₂O Molecular Weight: 146,1460 g/mol Schmelzpunkt: 216…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407363 CAS-Nummer: 4649-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...