6-Azaindole-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR46105-250MG OR46105-1G
APOSOR46105-250MGEA 54.1 EUR
APOSOR46105-250MG APOSOR46105-1G
6-Azaindole-2-carboxylic acid
6-Azaindole-2-carboxylic acid
Formel: C₈H₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11506121
CAS-Nummer: 24334-20-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR