6-(Dimethylamino)nicotinaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR46111-1G
APOSOR46111-1GEA
0
EUR
APOSOR46111-1G
6-(Dimethylamino)nicotinaldehyde
6-(Dimethylamino)nicotinaldehyde
Formel:
C₈H₁₀N₂O Schmelzpunkt: 51…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08272100 CAS-Nummer: 149805-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...