2-(Boc-amino)pyridine-5-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl (5-formylpyridin-2-yl)carbamate, 6-Aminonicotinaldehyde, 6-BOC protected

OR46112-1G
APOSOR46112-1GEA 184 EUR
APOSOR46112-1G
2-(Boc-amino)pyridine-5-carboxaldehyde
2-(Boc-amino)pyridine-5-carboxaldehyde
Formel: C₁₁H₁₄N₂O₃
Molecular Weight: 222,25 g/mol
Schmelzpunkt: 174…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08064228
CAS-Nummer: 199296-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR