5-Methoxypyridin-3-ol
Lieferant: Apollo Scientific
|
Achtung
|
OR46115-5G
APOSOR46115-5GEA
85.7
EUR
APOSOR46115-5G
5-Methoxypyridin-3-ol
5-Methoxypyridin-3-ol
Formel:
C6H7NO2 Schmelzpunkt: 147,5…148.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797672 CAS-Nummer: 109345-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...