(1R,2R)-1,2-Bis(methanesulphonyloxymethyl)cyclohexane

Lieferant: Apollo Scientific

OR46127-25G
APOSOR46127-25GEA 166 EUR
APOSOR46127-25G
(1R,2R)-1,2-Bis(methanesulphonyloxymethyl)cyclohexane
(1R,2R)-1,2-Bis(methanesulphonyloxymethyl)cyclohexane
Formel: C₁₀H₂₀O₆S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12923309
CAS-Nummer: 186204-35-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR