(3aR,7aR)-4'-(1,2-Benzisothiazol-3-yl) octahydro-spiro[2H-isoindole-2,1'-piperazinium] methanesulphonate

Lieferant: Apollo Scientific

Synonyme: (3aR,7aR)-4'-(Benzo[d]isothiazol-3-yl)octahydrospiro[isoindole-2,1'-piperazin]-2-ium methanesulphonate

OR46128-25G
APOSOR46128-25GEA 134 EUR
APOSOR46128-25G
(3aR,7aR)-4'-(1,2-Benzisothiazol-3-yl) octahydro-spiro[2H-isoindole-2,1'-piperazinium] methanesulphonate
(3aR,7aR)-4'-(1,2-Benzisothiazol-3-yl) octahydro-spiro[2H-isoindole-2,1'-piperazinium] methanesulphonate
Formel: C₁₉H₂₆N₄O₃S₂
Molecular Weight: 422,565 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28125607
CAS-Nummer: 186204-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR