cis-5-Norbornen-exo-2,3-dicarbonsäureanhydrid 98%
Lieferant: Apollo Scientific
Synonyme:
(3aR,4R,7S,7aS)-rel-3a,4,7,7a-Tetrahydro-4,7-methanoisobenzofuran-1,3-dione, Bicyclo[2.2.1]hept-5-ene-exo-2,3-dicarboxylic Anhydride
OR46134-100G
APOSOR46134-100GEA
179
EUR
APOSOR46134-100G
cis-5-Norbornen-exo-2,3-dicarbonsäureanhydrid 98%
cis-5-Norbornene exo-2,3-dicarboxylic anhydride
Formel:
C₉H₈O₃ Molecular Weight: 164,16 g/mol Schmelzpunkt: 140…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00630580 CAS-Nummer: 2746-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...