4-Hydroxy-3-iodobenzonitrile 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyano-2-iodophenol

OR46135-250MG
APOSOR46135-250MGEA 63.8 EUR
APOSOR46135-250MG
4-Hydroxy-3-iodobenzonitrile 97+%
4-Hydroxy-3-iodobenzonitrile
Formel: C₇H₄INO
Molecular Weight: 245,017 g/mol
Schmelzpunkt: 144…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11054826
CAS-Nummer: 2296-23-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR