5-Bromo-6-azaindole 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-1H-pyrrolo[2,3-c]pyridine
OR46136-5G
APOSOR46136-5GEA
127
EUR
APOSOR46136-5G
5-Bromo-6-azaindole 98%
5-Bromo-6-azaindole
Formel:
C₇H₅BrN₂ Molecular Weight: 197.032 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12024306 CAS-Nummer: 1215387-58-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...