2,6-Diiodopyrazine

Lieferant: Apollo Scientific

OR46149-5G
APOSOR46149-5GEA 212 EUR
APOSOR46149-5G
2,6-Diiodopyrazine
2,6-Diiodopyrazine
Formel: C₄H₂I₂N₂
Lagertemperatur: Kühlschrank
MDL: MFCD09265492
CAS-Nummer: 58138-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR