4-Amino-3-brom-2-chlorpyridin
Lieferant: Apollo Scientific
Synonyme:
3-Brom-2-chlor-4-pyridylamin
OR46206-25G
OR46206-5G
APOSOR46206-25GEA
473
EUR
APOSOR46206-25G
APOSOR46206-5G
4-Amino-3-brom-2-chlorpyridin
4-Amino-3-bromo-2-chloropyridine
Formel:
C₅H₄BrClN₂ Schmelzpunkt: 148…153 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09263478 CAS-Nummer: 215364-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...