1,4-Dibutoxynaphthalene-2,3-dicarbonitrile 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,4-Dibutoxy-2,3-dicyanonaphthalene

OR46229-1G
APOSOR46229-1GEA 209 EUR
APOSOR46229-1G
1,4-Dibutoxynaphthalene-2,3-dicarbonitrile 99%
1,4-Dibutoxynaphthalene-2,3-dicarbonitrile
Formel: C₂₀H₂₂N₂O₂
Molecular Weight: 322,401 g/mol
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191997
CAS-Nummer: 116453-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR