2,2'-Dibromacetophenon 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromophenacyl bromide, 2-Bromo-1-(2-bromophenyl)ethanone

OR46231-5G
APOSOR46231-5GEA 0 EUR
APOSOR46231-5G
2,2'-Dibromacetophenon 97%
2,2'-Dibromacetophenon
Formel: C₈H₆Br₂O
Molecular Weight: 277.9406 g/mol
Siedepunkt: 90…92 °C/0.02mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00044660
CAS-Nummer: 49851-55-0
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR