2,4-Dichloropyrimidine-5-carboxylic acid amide 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dichloropyrimidine-5-carboxamide

OR46232-5G
APOSOR46232-5GEA 89.9 EUR
APOSOR46232-5G
2,4-Dichloropyrimidine-5-carboxylic acid amide 95%
2,4-Dichloropyrimidine-5-carboxylic acid amide
Formel: C₅H₃Cl₂N₃O
Schmelzpunkt: 195 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD16658133
CAS-Nummer: 1240390-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR