Ethyl 2-(chloromethyl)-1,3-thiazole-4-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Chloromethyl)-4-(ethoxycarbonyl)-1,3-thiazole

OR46233-500MG
APOSOR46233-500MGEA 43.5 EUR
APOSOR46233-500MG
Ethyl 2-(chloromethyl)-1,3-thiazole-4-carboxylate 98%
Ethyl 2-(chloromethyl)-1,3-thiazole-4-carboxylate
Formel: C₇H₈ClNO₂S
Molecular Weight: 205,662 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06761633
CAS-Nummer: 842130-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR