4-Hydroxy-3-methoxybenzeneboronic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Borono-2-methoxyphenol, 5-Borono-2-hydroxyanisole, (4-Hydroxy-3-methoxyphenyl)boronic acid

OR46236-250MG
APOSOR46236-250MGEA 140 EUR
APOSOR46236-250MG
4-Hydroxy-3-methoxybenzeneboronic acid 95+%
4-Hydroxy-3-methoxybenzeneboronic acid
Formel: C₇H₉BO₄
Lagertemperatur: Gefrierschrank
MDL: MFCD18397223
CAS-Nummer: 182344-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR