1,5-Dimethyl-1H-pyrazol-3-ylamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,5-Dimethyl-1H-pyrazol-3-amine, 3-Amino-1,5-dimethyl-1H-pyrazole
OR46245-5G
APOSOR46245-5GEA
157
EUR
APOSOR46245-5G
1,5-Dimethyl-1H-pyrazol-3-ylamine
1,5-Dimethyl-1H-pyrazol-3-ylamine
Formel:
C₅H₉N₃ Molecular Weight: 111.15 g/mol Siedepunkt: 98 °C/3mmHg Schmelzpunkt: 65…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970041 CAS-Nummer: 35100-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...