4-Phenyl-1-piperidin-4-yl-1,3-dihydro-2H-imidazol-2-one 97%

Lieferant: Apollo Scientific
Achtung

OR46340-1G
APOSOR46340-1GEA 221 EUR
APOSOR46340-1G
4-Phenyl-1-piperidin-4-yl-1,3-dihydro-2H-imidazol-2-one 97%
4-Phenyl-1-piperidin-4-yl-1,3-dihydro-2H-imidazol-2-one
Formel: C₁₄H₁₇N₃O
Schmelzpunkt: 235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08061015
CAS-Nummer: 205058-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR