Methyl-3-amino-2-benzo[b]thiophencarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-aminobenzo[b]thiophene-2-carboxylate
OR4639-5G
APOSOR4639-5GEA
63.8
EUR
APOSOR4639-5G
Methyl-3-amino-2-benzo[b]thiophencarboxylat
Methyl-3-amino-2-benzo[b]thiophencarboxylat
Formel:
C₁₀H₉NO₂S Molecular Weight: 207,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00206744 CAS-Nummer: 35212-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...