6-Bromo-1H-imidazo[4,5-c]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-5-azabenzimidazole

OR46517-1G
APOSOR46517-1GEA 1320 EUR
APOSOR46517-1G
6-Bromo-1H-imidazo[4,5-c]pyridine
6-Bromo-1H-imidazo[4,5-c]pyridine
Formel: C₆H₄BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD19288874
CAS-Nummer: 1312440-90-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR