6-Nitropyridin-2-amine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-6-nitropyridine

OR46518-1G
APOSOR46518-1GEA 78.3 EUR
APOSOR46518-1G
6-Nitropyridin-2-amine 98%
6-Nitropyridin-2-amine
Formel: C₅H₅N₃O₂
Molecular Weight: 139.1121 g/mol
Schmelzpunkt: 160…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114202
CAS-Nummer: 14916-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR