Methyl 2-bromo-5-nitroisonicotinate 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-4-(methoxycarbonyl)-5-nitropyridine, Methyl 2-bromo-5-nitropyridine-4-carboxylate

OR46523-250MG
APOSOR46523-250MGEA 72.5 EUR
APOSOR46523-250MG
Methyl 2-bromo-5-nitroisonicotinate 99%
Methyl 2-bromo-5-nitroisonicotinate
Formel: C₇H₅BrN₂O₄
Molecular Weight: 261,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21606315
CAS-Nummer: 1361385-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR