Methyl-6-hydroxy-5-nitronicotinat
Lieferant: Apollo Scientific
|
Gefahr
|
OR46526-1G
APOSOR46526-1GEA
34.8
EUR
APOSOR46526-1G
Methyl-6-hydroxy-5-nitronicotinat
Methyl-6-hydroxy-5-nitronicotinat
Formel:
C₇H₆N₂O₅ Schmelzpunkt: 214…216 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699641 CAS-Nummer: 222970-61-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...