2-Chloro-6-methylnicotinic acid methyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-chloro-6-methylnicotinate

OR46529-1G
APOSOR46529-1GEA 49.3 EUR
APOSOR46529-1G
2-Chloro-6-methylnicotinic acid methyl ester
2-Chloro-6-methylnicotinic acid methyl ester
Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845763
CAS-Nummer: 53277-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR