Benzimidazole-6-carbonitrile 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1H-Benzo[d]imidazole-6-carbonitrile

OR46542-5G
APOSOR46542-5GEA 144 EUR
APOSOR46542-5G
Benzimidazole-6-carbonitrile 98%
Benzimidazole-6-carbonitrile
Formel: C₈H₅N₃
Molecular Weight: 143,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06809988
CAS-Nummer: 6287-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR