1-(Bromomethyl)-4-ethoxybenzene
Lieferant: Apollo Scientific
|
Gefahr
|
OR46549-1G
APOSOR46549-1GEA
783
EUR
APOSOR46549-1G
1-(Bromomethyl)-4-ethoxybenzene
1-(Bromomethyl)-4-ethoxybenzene
Formel:
C₉H₁₁BrO Siedepunkt: 134…136 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD08444317 CAS-Nummer: 2606-57-7 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...