2-Amino-5-nitropyrimidine-4,6-diol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4,6-dihydroxy-5-nitro-1,3-diazine, 2-Amino-4,6-dihydroxy-5-nitropyrimidine
OR4656-250MG
APOSOR4656-250MGEA
102
EUR
APOSOR4656-250MG
2-Amino-5-nitropyrimidine-4,6-diol 95%
2-Amino-5-nitropyrimidine-4,6-diol
Formel:
C₄H₄N₄O₄ Molecular Weight: 172,099 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082801 CAS-Nummer: 80466-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...