Bis(benzonitril)dichlorpalladium(II) 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Bis(benzonitrile)palladium chloride, Dichlorobis(benzonitrile)palladium(II), Bis(benzonitrile)palladium(II) Dichloride

OR46569-1G
APOSOR46569-1GEA 90.2 EUR
APOSOR46569-1G
Bis(benzonitril)dichlorpalladium(II) 95+%
Bis(benzonitril)dichlorpalladium(II)
Formel: C₁₄H₁₀Cl₂N₂Pd
Molecular Weight: 383,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013123
CAS-Nummer: 14220-64-5
EINECS: 238-085-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR