7-Chinolinamin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Quinolin-7-amine
OR46576-1G
APOSOR46576-1GEA
24.8
EUR
APOSOR46576-1G
7-Chinolinamin
7-Aminoquinoline
Formel:
C₉H₈N₂ Molecular Weight: 144,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646243 CAS-Nummer: 580-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...