4-Chloro-1H-imidazole 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-1H-imidazole

OR46577-1G OR46577-5G
APOSOR46577-1GEA 62.5 EUR
APOSOR46577-1G APOSOR46577-5G
4-Chloro-1H-imidazole 98%
4-Chloro-1H-imidazole
Formel: C₃H₃ClN₂
Molecular Weight: 102.52 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09033087
CAS-Nummer: 15965-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR