Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate 95+%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-(1,4-dioxaspiro[4.5]decan-8-ylidene)acetate

OR46595-1G
APOSOR46595-1GEA 119 EUR
APOSOR46595-1G
Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate 95+%
Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate
Formel: C₁₂H₁₈O₄
Molecular Weight: 226,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779434
CAS-Nummer: 51656-91-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR