Bis(2-aminophenyl)sulphide

Lieferant: Apollo Scientific
Achtung

OR4660-25G OR4660-5G
APOSOR4660-25GEA 111 EUR
APOSOR4660-25G APOSOR4660-5G
Bis(2-aminophenyl)sulphide
Bis(2-aminophenyl)sulphide
Formel: C₁₂H₁₂N₂S
Schmelzpunkt: 85…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094662
CAS-Nummer: 5873-51-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR