(2S)-2-Aminobutanamidhydrochlorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-Abu-NH2 · HCl, (2S)-2-Aminobutanamide hydrochloride, (S)-(+)-2-Aminobutanamide hydrochloride, H-Abu-NH2 HCl
OR46608-100G
APOSOR46608-100GEA
68.3
EUR
APOSOR46608-100G
(2S)-2-Aminobutanamidhydrochlorid 97%
(S)-2-Aminobutanamide hydrochloride
Formel:
C₄H₁₁ClN₂O Molecular Weight: 138,60 g/mol Schmelzpunkt: 259…263 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00136565 CAS-Nummer: 7682-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...