3-Phenyl-1H-pyrazol-4-amine
Lieferant: Apollo Scientific
OR46626-1G
APOSOR46626-1GEA
246
EUR
APOSOR46626-1G
3-Phenyl-1H-pyrazol-4-amine
3-Phenyl-1H-pyrazol-4-amine
Formel:
C₉H₉N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01693716 CAS-Nummer: 91857-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...