N-Cyclobutyl-N-cyclopropylamine

Lieferant: Apollo Scientific

OR46631-1G
APOSOR46631-1GEA 394 EUR
APOSOR46631-1G
N-Cyclobutyl-N-cyclopropylamine
N-Cyclobutyl-N-cyclopropylamine
Formel: C₇H₁₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD12963753
CAS-Nummer: 823-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR