4-Methyl-1H-pyrrole-2-carbaldehyde

Lieferant: Apollo Scientific

OR46634-1G
APOSOR46634-1GEA 886 EUR
APOSOR46634-1G
4-Methyl-1H-pyrrole-2-carbaldehyde
4-Methyl-1H-pyrrole-2-carbaldehyde
Formel: C₆H₇NO
Molecular Weight: 109,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08235196
CAS-Nummer: 24014-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR