[1-(3-Methoxyphenyl)cyclopropyl]methylamine

Lieferant: Apollo Scientific

OR46641-1G
APOSOR46641-1GEA 432 EUR
APOSOR46641-1G
[1-(3-Methoxyphenyl)cyclopropyl]methylamine
[1-(3-Methoxyphenyl)cyclopropyl]methylamine
Formel: C₁₁H₁₅NO
Lagertemperatur: Kühlschrank
MDL: MFCD07374430
CAS-Nummer: 886365-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR