N-Methyl-N-[(4-methylpyridin-2-yl)methyl]amine

Lieferant: Apollo Scientific

OR46642-1G
APOSOR46642-1GEA 429 EUR
APOSOR46642-1G
N-Methyl-N-[(4-methylpyridin-2-yl)methyl]amine
N-Methyl-N-[(4-methylpyridin-2-yl)methyl]amine
Formel: C₈H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800577
CAS-Nummer: 1060804-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR