3-(4-Methylphenyl)oxirane-2-carboxylic acid

Lieferant: Apollo Scientific

OR46651-1G
APOSOR46651-1GEA 667 EUR
APOSOR46651-1G
3-(4-Methylphenyl)oxirane-2-carboxylic acid
3-(4-Methylphenyl)oxirane-2-carboxylic acid
Formel: C₁₀H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18089207
CAS-Nummer: 736091-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR