4-Bromo-3,5-dimethyl-1H-pyrrole-2-carboxylic acid

Lieferant: Apollo Scientific

OR46656-1G
APOSOR46656-1GEA 516 EUR
APOSOR46656-1G
4-Bromo-3,5-dimethyl-1H-pyrrole-2-carboxylic acid
4-Bromo-3,5-dimethyl-1H-pyrrole-2-carboxylic acid
Formel: C₇H₈BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11557240
CAS-Nummer: 562074-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR