5-Bromo-3,4-dimethyl-1H-pyrrole-2-carboxylic acid

Lieferant: Apollo Scientific

OR46657-1G
APOSOR46657-1GEA 2190 EUR
APOSOR46657-1G
5-Bromo-3,4-dimethyl-1H-pyrrole-2-carboxylic acid
5-Bromo-3,4-dimethyl-1H-pyrrole-2-carboxylic acid
Formel: C₇H₈BrNO₂
Molecular Weight: 218,048 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13705413
CAS-Nummer: 861034-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR