(2,6-Dimethyltetrahydro-2H-pyran-4-yl)acetic acid
Lieferant: Apollo Scientific
OR46658-1G
APOSOR46658-1GEA
545
EUR
APOSOR46658-1G
(2,6-Dimethyltetrahydro-2H-pyran-4-yl)acetic acid
(2,6-Dimethyltetrahydro-2H-pyran-4-yl)acetic acid
Formel:
C₉H₁₆O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100081 CAS-Nummer: 1215417-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...