2-(2-Oxocyclohexyl)acetic acid

Lieferant: Apollo Scientific

OR46662-1G
APOSOR46662-1GEA 451 EUR
APOSOR46662-1G
2-(2-Oxocyclohexyl)acetic acid
2-(2-Oxocyclohexyl)acetic acid
Formel: C₈H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD20638190
CAS-Nummer: 1438-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR