4-Chloro-3-oxobutanoic acid

Lieferant: Apollo Scientific
Gefahr

OR46667-1G
APOSOR46667-1GEA 667 EUR
APOSOR46667-1G
4-Chloro-3-oxobutanoic acid
4-Chloro-3-oxobutanoic acid
Formel: C₄H₅ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00129128
CAS-Nummer: 27807-84-7
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR