Benzyltriethylammoniumchlorid 99%
Lieferant: Apollo Scientific
|
Achtung
|
OR4667-500G
OR4667-5KG
OR4667-1KG
APOSOR4667-500GEA
24.8
EUR
APOSOR4667-500G
APOSOR4667-5KG
APOSOR4667-1KG
Benzyltriethylammoniumchlorid 99%
Benzyltriethylammoniumchlorid
Formel:
C₁₃H₂₂ClN Siedepunkt: 444,8 °C Flammpunkt: >275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011824 CAS-Nummer: 56-37-1 EINECS: 200-270-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...