2-Chloro-4-(methylsulphonyl)benzoyl chloride

Lieferant: Apollo Scientific

OR46672-1G OR46672-250MG OR46672-500MG OR46672-2.5G OR46672-5G
APOSOR46672-1GEA 432 EUR
APOSOR46672-1G APOSOR46672-250MG APOSOR46672-500MG APOSOR46672-2.5G APOSOR46672-5G
2-Chloro-4-(methylsulphonyl)benzoyl chloride
2-Chloro-4-(methylsulphonyl)benzoyl chloride
Formel: C₈H₆Cl₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD12025154
CAS-Nummer: 106904-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR