4-(4-Methylpiperazin-1-yl)benzoyl chloride

Lieferant: Apollo Scientific

OR46673-1G OR46673-100MG OR46673-250MG OR46673-500MG OR46673-5G
APOSOR46673-1GEA 0 EUR
APOSOR46673-1G APOSOR46673-100MG APOSOR46673-250MG APOSOR46673-500MG APOSOR46673-5G
4-(4-Methylpiperazin-1-yl)benzoyl chloride
4-(4-Methylpiperazin-1-yl)benzoyl chloride
Formel: C₁₂H₁₅ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD20483877
CAS-Nummer: 401495-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR